missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG H and L Chain Goat anti-Rabbit, Unlabeled, Polyclonal, MP Biomedicals
Goat Polyclonal Antibody
Brand: MP Biomedicals 08610032
Additional Details : Weight : 0.00500kg
Description
IgG H and L Chain Polyclonal antibody specifically detects IgG H and L Chain in Rabbit samples. It is validated for Immunohistochemistry.Specifications
IgG H and L Chain | |
Polyclonal | |
Goat | |
H and L Chain | |
Rabbit | |
Purified |
Immunohistochemistry | |
Unconjugated | |
5 mL | |
Secondary | |
Store at -20°C. |