missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-949 Microfinished Nitrile-Coated Gloves with Intercept Technology™
GREENER_CHOICE

Catalog Number p-7215033
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Microfinished nitrile coating enhances grip capabilities while also resisting residual oils and grease. Intercept Technology™ liner provides ANSI Cut Level 2 Protection.

  • Offers cut, abrasion and oil resistance in various dip geometries
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology increases abrasion resistance
  • Microfinished nitrile coating enhances grip capabilities
  • Coating is resistant to residual oils and greases present on handled parts
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Hand Specific
Material Intercept Technology Yarn
Size 7
EN388 Standard 4242B
For Use With (Application) Maintenance, Automotive Assembly, Metal Fabrication, and Glass Manufacturing
Glove Cuff Color Black
Glove Cuff Style Knit
Series 11-949
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'