Learn More
Ansell HyFlex™ 11-725 Polyurethane-Coated Cut-Resistant Lycra™ Gloves
Gloves combine comfort and dexterity with a high degree of cut resistance for applications that involve sharp objects.
Brand:  Ansell 11725070
Additional Details : Weight : 5.32800kg
 View more versions of this product
Product Code. 16649073
  • 127.00 € / 12 pairs
Estimated Shipment: 03-01-2022
Add to basket
Description
Description
  • A balance of comfort, dexterity, and sure handling
  • High level of cut resistance for safe use in applications involving contact with sharp piece
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Lycra™ in the liner/coating provides enhances dexterity and fit
  • Designed for use in dry, as well as slightly oily environments
Specifications
Specifications
White
Knit
11-725
EN 420:2003 + A1:2009, REACH Compliant
Polyurethane
15
Automotive Assembly, Construction, Maintenance, and Metal Fabrication
Ansell Intercept Technology™ Yarn
7
4342B
Specific