missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-100 Nylon Industrial Gloves with Ionic+™ Antimicrobial Technology

Ionic+™ is an EPA-registered antimicrobial technology that leverages the antimicrobial power of silver to protect gloves from microbial impacts. Gloves are protected from bacterial-related odors, stains, and fabric degradation.

Brand:  Ansell 11100110

Additional Details : Weight : 8.73000kg

 View more versions of this product

Product Code. 16099881

  • 54.50 € / 12 pairs


Estimated Shipment
18-06-2021
Add to basket

Description

Description

  • Industrial gloves designed with Ionic+™, an EPA-registered antimicrobial technology
  • Ionic+™ coating is permanent, and lasts for the glove's entire life
  • Antimicrobial power of silver protects gloves from microbial impacts
  • All day comfort with high dexterity in a light-weight breathable design
  • Self-Sanitizing 360° antimicrobial protection of both the liner and coating
  • Compliant with EU Biocides Regulation 528/2012 for treated articles
Specifications

Specifications

Gray
Knit
Lined
Coated
11
ANSI 3
Nitrile
Specific
Unisex
Ionic+™ Active
Nylon
11-100
EN 420:2003 + A1:2009, REACH Compliant
2131A
ANSI A1
15
Videos
SDS
Documents