missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum, New Zealand Origin

Produced using true pool technology for uniformity and consistency from bottle to bottle

Brand:  Cytiva SH30406.01IR25-40

Additional Details : Weight : 0.14000kg

 View more versions of this product

Product Code. 16655755

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Sourced in New Zealand with complete traceability back to the original source
  • Filtered through three sequential 100 nm pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
Specifications

Specifications

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase, Glutamic Oxaloacetic Transaminase, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, Total Iron Binding Capacity (TIBC), Percent Iron Saturation, Glucose, and IgG
≤-10°C
New Zealand
Fetal Bovine Serum
Sterile
Irradiated (25-40 kGy)
100 mL
Enhancing Cell Growth
<25 mg/dL
USDA Approved
<10 days
Serum
100 mL
Sequential Triple-filtered, 100 nm
Virus Panel Testing According to 9 CFR 113.53
≤ 20 EU/mL
No
HyClone™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions