missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, New Zealand Origin
Produced using true pool technology for uniformity and consistency from bottle to bottle
255.00 €
Specifications
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase, Glutamic Oxaloacetic Transaminase, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, Total Iron Binding Capacity (TIBC), Percent Iron Saturation, Glucose, and IgG |
---|---|
Certifications/Compliance | USDA Approved |
Content And Storage | ≤-10°C |
Age | <10 days |
Country of Origin | New Zealand |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
16645755
|
Cytiva
SH30406.01HI |
Heat Inactivated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16655755
|
Cytiva
SH30406.01IR25-40 |
Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Sourced in New Zealand with complete traceability back to the original source
- Filtered through three sequential 100 nm pore size-rated filters
- Virus panel testing according to 9 CFR 113.53
- Low in antibodies and high in growth factors
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase, Glutamic Oxaloacetic Transaminase, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, Total Iron Binding Capacity (TIBC), Percent Iron Saturation, Glucose, and IgG | |
≤-10°C | |
New Zealand | |
Fetal Bovine Serum | |
Sterile | |
Virus Panel Testing According to 9 CFR 113.53 | |
≤ 20 EU/mL | |
<25 mg/dL |
USDA Approved | |
<10 days | |
Serum | |
100 mL | |
Sequential Triple-filtered, 100 nm | |
100 mL | |
Enhancing Cell Growth | |
HyClone™ |