missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF364 Full-length ORF (NP_055270.1, 1 a.a. - 304 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027246-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138702

  • 335.00 € / 10µg
Estimated Shipment: 21-08-2024
to see stock.

Description

Description

Sequence: MAEASAAGADSGAAVAAHRFFCHFCKGEVSPKLPEYICPRCESGFIEEVTDDSSFLGGGGSRIDNTTTTHFAELWGHLDHTMFFQDFRPFLSSSPLDQDNRANERGHQTHTDFWGARPPRLPLGRRYRSRGSSRPDRSPAIEGILQHIFAGFFANSAIPGSPHPFSWSGMLHSNPGDYAWGQTGLDAIVTQLLGQLENTGPPPADKEKITSLPTVTVTQEQVDMGLECPVCKEDYTVEEEVRQLPCNHFFHSSCIVPWLELHDTCPVCRKSLNGEDSTRQSQSTEASASNRFSNDSQLHDRWTF
Specifications

Specifications

NP_055270.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCA2/ZNF364
RNF115
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
27246
ZNF364 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEASAAGADSGAAVAAHRFFCHFCKGEVSPKLPEYICPRCESGFIEEVTDDSSFLGGGGSRIDNTTTTHFAELWGHLDHTMFFQDFRPFLSSSPLDQDNRANERGHQTHTDFWGARPPRLPLGRRYRSRGSSRPDRSPAIEGILQHIFAGFFANSAIPGSPHPFSWSGMLHSNPGDYAWGQTGLDAIVTQLLGQLENTGPPPADKEKITSLPTVTVTQEQVDMGLECPVCKEDYTVEEEVRQLPCNHFFHSSCIVPWLELHDTCPVCRKSLNGEDSTRQSQSTEASASNRFSNDSQLHDRWTF
RUO
RNF115
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions