missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF254 Full-length ORF (AAH43147.2, 1 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009534-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16183802

  • 508.00 € / 25µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]

Sequence: MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV
Specifications

Specifications

AAH43147.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
103.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BMZF-5/FLJ58216/HD-ZNF1/ZNF539/ZNF91L
ZNF254
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9534
ZNF254 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV
RUO
ZNF254
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions