missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF254 Full-length ORF (AAH43147.2, 1 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH43147.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9534 |
Molecular Weight (g/mol) | 103.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16183802
|
Abnova™
H00009534-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16173802
|
Abnova™
H00009534-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]
Sequence: MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQVSpecifications
AAH43147.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
103.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV | |
RUO | |
ZNF254 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9534 | |
ZNF254 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMZF-5/FLJ58216/HD-ZNF1/ZNF539/ZNF91L | |
ZNF254 | |
Recombinant | |
wheat germ expression system |