missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VPS16 Partial ORF (NP_072097.2, 754 a.a. - 839 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00064601-Q01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16139686

  • 302.00 € / 10 Mikrogramm


Verfügbar ab
24-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: IGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK
Spezifikation

Spezifikation

NP_072097.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.2kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK
RUO
VPS16
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
64601
VPS16 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
hVPS16
VPS16
Yes
Videos
Dokumentation