missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UNC13D Partial ORF (NP_954712.1, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_954712.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 201294 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055815
|
Abnova™
H00201294-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045815
|
Abnova™
H00201294-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder. [provided by RefSeq]
Sequence: ATLLSHPQQRPPFLRQAIKIRRRRVRDLQDPPPQMAPEIQPPSHHFSPEQRALLYEDALYTVLHRLGHPEPNHVTEASELLRYLQEAFHVEPEEHQQTLSpecifications
NP_954712.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FHL3/HLH3/HPLH3/Munc13-4 | |
UNC13D | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
201294 | |
UNC13D (Human) Recombinant Protein (Q01) | |
ATLLSHPQQRPPFLRQAIKIRRRRVRDLQDPPPQMAPEIQPPSHHFSPEQRALLYEDALYTVLHRLGHPEPNHVTEASELLRYLQEAFHVEPEEHQQTL | |
RUO | |
UNC13D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |