missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSKS Partial ORF (NP_068379, 471 a.a. - 560 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00060385-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139386

  • 508.00 € / 25µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression. [provided by RefSeq]

Sequence: ALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDH
Specifications

Specifications

NP_068379
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDH
RUO
TSKS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
60385
TSKS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TSKS1
TSKS
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions