missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSGA2 Partial ORF (NP_543136, 200 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00089765-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101057

  • 518.16 € / 25µg

Estimated Shipment: 14-01-2022
Add to basket

Description

Description

Sequence: EELVTVVPKWKATQITELALWTPTLPKKPTSTDGPGQDAPGAESAGEPGEEAQALLEGFEGEMDMRPGDEDADVLREESREYDQEEFRYDMDEGNINSEEEETRQSDLQD
Specifications

Specifications

NP_543136
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EELVTVVPKWKATQITELALWTPTLPKKPTSTDGPGQDAPGAESAGEPGEEAQALLEGFEGEMDMRPGDEDADVLREESREYDQEEFRYDMDEGNINSEEEETRQSDLQD
RUO
RSPH1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
89765
TSGA2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126568/MGC141927/RSP44/RSPH10A/TSA2/TSGA2
RSPH1
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions