Learn More
Abnova™ Human TOPBP1 Partial ORF (NP_008958, 1327 a.a. - 1435 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011073-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a binding protein which interacts with the C-terminal region of topoisomerase II beta. This interaction suggests a supportive role for this protein in the catalytic reactions of topoisomerase II beta through transient breakages of DNA strands. [provided by RefSeq]
Sequence: LLQSGGAKVLPGHSVPLFKEATHLFSDLNKLKPDDSGVNIAEAAAQNVYCLRTEYIADYLMQESPPHVENYCLPEAISFIQNNKELGTGLSQKRKAPTEKNKIKRPRVHSpecifications
NP_008958 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LLQSGGAKVLPGHSVPLFKEATHLFSDLNKLKPDDSGVNIAEAAAQNVYCLRTEYIADYLMQESPPHVENYCLPEAISFIQNNKELGTGLSQKRKAPTEKNKIKRPRVH | |
RUO | |
TOPBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11073 | |
TOPBP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TOP2BP1 | |
TOPBP1 | |
Recombinant | |
wheat germ expression system |