missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TIMM50 Partial ORF (NP_001001563, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00092609-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101237

  • 341.70 € / 10µg

Estimated Shipment: 24-01-2022
Add to basket

Description

Description

Sequence: GMTAFPLIDSVDPHGFISYRLFRDATRYMDGHHVKDISCLNRDPARVVVVDCKKEAFRLQPYNGVALRPWDGNSDDRVLLDLSAFLKTIALNGVEDVRTV
Specifications

Specifications

NP_001001563
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GMTAFPLIDSVDPHGFISYRLFRDATRYMDGHHVKDISCLNRDPARVVVVDCKKEAFRLQPYNGVALRPWDGNSDDRVLLDLSAFLKTIALNGVEDVRTV
RUO
TIMM50
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
92609
TIMM50 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC102733/TIM50/TIM50L
TIMM50
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions