missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TIMELESS Partial ORF (NP_003911, 1109 a.a. - 1207 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008914-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139645

  • 508.00 € / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: PELQPKVPGEQGSDEEHCKEHRAQALRALLLAHKKKAGLASPEEEDAVGKEPLKAAPKKRQLLDSDEEQEEDEGRNRAPELGAPGIQKKKRYQIEDDED
Specifications

Specifications

NP_003911
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PELQPKVPGEQGSDEEHCKEHRAQALRALLLAHKKKAGLASPEEEDAVGKEPLKAAPKKRQLLDSDEEQEEDEGRNRAPELGAPGIQKKKRYQIEDDED
RUO
TIMELESS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8914
TIMELESS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12640/FLJ20714/TIM/TIM1/hTIM
TIMELESS
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions