missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TCF3 Partial ORF (NP_003191, 545 a.a. - 654 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003191 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6929 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16186345
|
Abnova™
H00006929-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16176345
|
Abnova™
H00006929-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TCF3 gene, also called E2A, encodes 2 basic helix-loop-helix (bHLH) transcription factors, E12 and E47, through alternative splicing. E12 and E47 are involved in regulation of immunoglobulin gene expression (Bain et al., 1994 [PubMed 8001125]).[supplied by OMIM]
Sequence: EREKERRVANNARERLRVRDINEAFKELGRMCQLHLNSEKPQTKLLILHQAVSVILNLEQQVRERNLNPKAACLKRREEEKVSGVVGDPQMVLSAPHPGLSEAHNPAGHMSpecifications
NP_003191 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E2A/ITF1/MGC129647/MGC129648/bHLHb21 | |
TCF3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6929 | |
TCF3 (Human) Recombinant Protein (Q01) | |
EREKERRVANNARERLRVRDINEAFKELGRMCQLHLNSEKPQTKLLILHQAVSVILNLEQQVRERNLNPKAACLKRREEEKVSGVVGDPQMVLSAPHPGLSEAHNPAGHM | |
RUO | |
TCF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |