missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ST8SIA5 Partial ORF (NP_037437, 33 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00029906-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: QQILYGRNYIKRYFEFYEGPFEYNSTRCLELRHEILEVKVLSMVKQSELFDRWKSLQMCKWAMNISEANQFKSTLSRCCNAPAFLFTTQKNTPLGTKLKYSpezifikation
NP_037437 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QQILYGRNYIKRYFEFYEGPFEYNSTRCLELRHEILEVKVLSMVKQSELFDRWKSLQMCKWAMNISEANQFKSTLSRCCNAPAFLFTTQKNTPLGTKLKY | |
RUO | |
ST8SIA5 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
29906 | |
ST8SIA5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119670|MGC119671|SIAT8E|ST8SiaV | |
ST8SIA5 | |
Yes |