missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SOX11 Full-length ORF (AAI60119.1, 1 a.a. - 441 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006664-P01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MVQQAESLEAESNLPREALDTEEGEFMACSPVALDESDPDWCKTASGHIKRPMNAFMVWSKIERRKIMEQSPDMHNAEISKRLGKRWKMLKDSEKIPFIREAERLRLKHMADYPDYKYRPRKKPKMDPSAKPSASQSPEKSAAGGGGGSAGGGAGGAKTSKGSSKKCGKLKAPAAAGAKAGAGKAAQSGDYGGAGDDYVLGSLRVSGSGGGGAGKTVKCVFLDEDDDDDDDDDELQLQIKQEPDEEDEEPPHQQLLQPPGQQPSQLLRRYNVAKVPASPTLSSSAESPEGASLYDEVRAGATSGAGGGSRLYYSFKNITKQHPPPLAQPALSPASSRSVSTSSSSSSGSSSGSSGEDADDLMFDLSLNFSQSAHSASEQQLGGGAAAGNLSLSLVDKDLDSFSEGSLGSHFEFPDYCTPELSEMIAGDWLEANFSDLVFTYSpezifikation
AAI60119.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SOX11 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
6664 | |
SOX11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MVQQAESLEAESNLPREALDTEEGEFMACSPVALDESDPDWCKTASGHIKRPMNAFMVWSKIERRKIMEQSPDMHNAEISKRLGKRWKMLKDSEKIPFIREAERLRLKHMADYPDYKYRPRKKPKMDPSAKPSASQSPEKSAAGGGGGSAGGGAGGAKTSKGSSKKCGKLKAPAAAGAKAGAGKAAQSGDYGGAGDDYVLGSLRVSGSGGGGAGKTVKCVFLDEDDDDDDDDDELQLQIKQEPDEEDEEPPHQQLLQPPGQQPSQLLRRYNVAKVPASPTLSSSAESPEGASLYDEVRAGATSGAGGGSRLYYSFKNITKQHPPPLAQPALSPASSRSVSTSSSSSSGSSSGSSGEDADDLMFDLSLNFSQSAHSASEQQLGGGAAAGNLSLSLVDKDLDSFSEGSLGSHFEFPDYCTPELSEMIAGDWLEANFSDLVFTY | |
RUO | |
SOX11 | |
Yes |