missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SOAT1 Partial ORF (NP_003092.4, 249 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006646-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16084735

  • 335.00 € / 10µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acyl-coenzyme A:cholesterol acyltransferase (ACACT; EC 2.3.1.26) is an intracellular protein located in the endoplasmic reticulum that forms cholesterol esters from cholesterol. Accumulation of cholesterol esters as cytoplasmic lipid droplets within macrophages and smooth muscle cells is a characteristic feature of the early stages of atherosclerotic plaques (Cadigan et al., 1988 [PubMed 3335499]).[supplied by OMIM]

Sequence: PPASRFIIIFEQIRFVMKAHSFVRENVPRVLNSAKEKSSTVPIPTVNQYLYFLFAPTLIYRDSYPRNPTVRWG
Specifications

Specifications

NP_003092.4
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PPASRFIIIFEQIRFVMKAHSFVRENVPRVLNSAKEKSSTVPIPTVNQYLYFLFAPTLIYRDSYPRNPTVRWG
RUO
SOAT1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6646
SOAT1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACACT/ACAT/ACAT1/RP11-215I23.2/SOAT/STAT
SOAT1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions