missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC25A6 Partial ORF (NP_001627, 135 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000293-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101601

  • 518.16 € / 25µg

Estimated Shipment: 21-01-2022
Add to basket

Description

Description

This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene. [provided by RefSeq]

Sequence: DFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKGMLPDPKNTH
Specifications

Specifications

NP_001627
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.99kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DFARTRLAADVGKSGTEREFRGLGDCLVKITKSDGIRGLYQGFSVSVQGIIIYRAAYFGVYDTAKGMLPDPKNTH
RUO
SLC25A6
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
293
SLC25A6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AAC3/ANT3/ANT3Y/MGC17525
SLC25A6
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions