missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SH3GL2 Partial ORF (NP_003017, 64 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003017 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6456 |
Molecular Weight (g/mol) | 32.45kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185215
|
Abnova™
H00006456-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175215
|
Abnova™
H00006456-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: SRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMRELSpecifications
NP_003017 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNSA2/EEN-B1/FLJ20276/FLJ25015/SH3D2A/SH3P4 | |
SH3GL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6456 | |
SH3GL2 (Human) Recombinant Protein (Q01) | |
SRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMREL | |
RUO | |
SH3GL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |