missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SH3GL2 Full-length ORF (NP_003017.1, 1 a.a. - 352 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006456-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MSVAGLKKQFHKATQKVSEKVGGAEGTKLDDDFKEMERKVDVTSRAVMEIMTKTIEYLQPNPASRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMRELSEVKDSLDIEVKQNFIDPLQNLHDKDLREIQHHLKKLEGRRLDFDYKKKRQGKIPDEELRQALEKFDESKEIAESSMFNLLEMDIEQVSQLSALVQAQLEYHKQAVQILQQVTVRLEERIRQASSQPRREYQPKPRMSLEFPTGDSTQPNGGLSHTGTPKPSGVQMDQPCCRALYDFEPENEGELGFKEGDIITLTNQIDENWYEGMLHGHSGFFPINYVEILVALPHSpecifications
NP_003017.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNSA2/EEN-B1/FLJ20276/FLJ25015/SH3D2A/SH3P4 | |
SH3GL2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6456 | |
SH3GL2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVAGLKKQFHKATQKVSEKVGGAEGTKLDDDFKEMERKVDVTSRAVMEIMTKTIEYLQPNPASRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMRELSEVKDSLDIEVKQNFIDPLQNLHDKDLREIQHHLKKLEGRRLDFDYKKKRQGKIPDEELRQALEKFDESKEIAESSMFNLLEMDIEQVSQLSALVQAQLEYHKQAVQILQQVTVRLEERIRQASSQPRREYQPKPRMSLEFPTGDSTQPNGGLSHTGTPKPSGVQMDQPCCRALYDFEPENEGELGFKEGDIITLTNQIDENWYEGMLHGHSGFFPINYVEILVALPH | |
RUO | |
SH3GL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |