missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SFXN3 Partial ORF (AAH00124.2, 125 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH00124.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81855 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075695
|
Abnova™
H00081855-Q02.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065695
|
Abnova™
H00081855-Q02.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: WVNQSFNAIVNYSNRSGDTPITVRQLGTAYVSATTGAVATALGLKSLTKHLPPLVGRFVPFAAVAAANCINIPLMRQRELQVGIPVADEAGQRLGYSVTASpecifications
AAH00124.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BA108L7.2/SFX3 | |
SFXN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
81855 | |
SFXN3 (Human) Recombinant Protein (Q02) | |
WVNQSFNAIVNYSNRSGDTPITVRQLGTAYVSATTGAVATALGLKSLTKHLPPLVGRFVPFAAVAAANCINIPLMRQRELQVGIPVADEAGQRLGYSVTA | |
RUO | |
SFXN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |