Learn More
Abnova™ Human SEC8L1 Partial ORF (NP_068579, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_068579 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 60412 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16149396
|
Abnova™
H00060412-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139396
|
Abnova™
H00060412-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MAAEAAGGKYRSTVSKSKDPSGLLISVIRTLSTSDDVEDRENEKGRLEEAYEKCDRDLDELIVQHYTELTTAIRTYQSITERITNSRNKIKQVKENLLSCKMLLHCKRDSpecifications
NP_068579 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC27170/REC8/SEC8/SEC8L1/Sec8p | |
EXOC4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
60412 | |
SEC8L1 (Human) Recombinant Protein (Q01) | |
MAAEAAGGKYRSTVSKSKDPSGLLISVIRTLSTSDDVEDRENEKGRLEEAYEKCDRDLDELIVQHYTELTTAIRTYQSITERITNSRNKIKQVKENLLSCKMLLHCKRD | |
RUO | |
EXOC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |