missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RRP15 Full-length ORF (AAH20641.1, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00051018-P01.25ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16139512

  • 458.00 € / 25 Mikrogramm


Verfügbar ab
28-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MKMVTGAVASVLEDEATDTSDSEGSCGSEKDHFYSDDDAIEADSEGDAEPCDKENENDGESSVGTNMGWADAMAKVLNKKTPESKPTILVKNKKLEKEKEKLKQERLEKIKQRDKRLEWEMMCRVKPDVVQDKETERNLQRIATRGVVQLFNAVQKHQKNVDEKVKEAGSSMRKRAKLISTVSKKDFISVLRGMDGSTNETASSRKKPKAKQTEVKSEEGPGWTILRDDFMMGASMKDWDKESDGPDDSRPESASDSDT
Spezifikation

Spezifikation

AAH20641.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CGI-115|KIAA0507|MGC22291
RRP15
Yes
wheat germ expression system
Liquid
51018
RRP15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKMVTGAVASVLEDEATDTSDSEGSCGSEKDHFYSDDDAIEADSEGDAEPCDKENENDGESSVGTNMGWADAMAKVLNKKTPESKPTILVKNKKLEKEKEKLKQERLEKIKQRDKRLEWEMMCRVKPDVVQDKETERNLQRIATRGVVQLFNAVQKHQKNVDEKVKEAGSSMRKRAKLISTVSKKDFISVLRGMDGSTNETASSRKKPKAKQTEVKSEEGPGWTILRDDFMMGASMKDWDKESDGPDDSRPESASDSDT
RUO
RRP15
Wheat Germ (in vitro)
GST
Videos
Dokumentation