missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF184 Partial ORF (NP_060603, 478 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055167-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137956

  • 335.00 € / 10µg
Estimated Shipment: 20-08-2024
to see stock.

Description

Description

Sequence: GQRCPCYSNRKACLDCICRGCQNSYMANGEKKLEAFAVPEKALEQTRLTLGINVTSIAVRNASTSTSVINVTGSPVTTFLAASTHDDKSLDEAIDMRF
Specifications

Specifications

NP_060603
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GQRCPCYSNRKACLDCICRGCQNSYMANGEKKLEAFAVPEKALEQTRLTLGINVTSIAVRNASTSTSVINVTGSPVTTFLAASTHDDKSLDEAIDMRF
RUO
MSL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55167
RNF184 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10546/KIAA1585/MSL-2/MSL2L1/RNF184
MSL2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions