Learn More
Abnova™ Human RNASE4 Full-length ORF (AAH15520, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006038-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene belongs to the pancreatic ribonuclease family. It plays an important role in mRNA cleavage and has marked specificity towards the 3' side of uridine nucleotides. Alternative splicing results in two transcript variants encoding the same protein. This gene and the gene that encodes angiogenin share promoters and 5' exons. Each gene splices to a unique downstream exon that contains its complete coding region. [provided by RefSeq]
Sequence: MALQRTHSLLLLLLLTLLGLGLVQPSYGQDGMYQRFLRQHVHPEETGGSDRHCNLMMQRRKMTLYHCKRFNTFIHEDIWNIRSICSTTNIQCKNGKMNCHEGVVKVTDCRDTGSSRAPNCRYRAIASTRRVVIACEGNPQVPVHFDGSpezifikation
AAH15520 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.91kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC9306|RNS4 | |
RNASE4 | |
Yes |
wheat germ expression system | |
Liquid | |
6038 | |
RNASE4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALQRTHSLLLLLLLTLLGLGLVQPSYGQDGMYQRFLRQHVHPEETGGSDRHCNLMMQRRKMTLYHCKRFNTFIHEDIWNIRSICSTTNIQCKNGKMNCHEGVVKVTDCRDTGSSRAPNCRYRAIASTRRVVIACEGNPQVPVHFDG | |
RUO | |
RNASE4 | |
Wheat Germ (in vitro) | |
GST |