missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RIPK1 Partial ORF (NP_003795, 562 a.a. - 671 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | RIPK1 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
Molecular Weight (g/mol) | 37.84kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
Sequence: TNFKEEPAAKYQAIFDNTTSLTDKHLDPIRENLGKHWKNCARKLGFTQSQIDEIDHDYERDGLKEKVYQMLQKWVMREGIKGATVGKLAQALHQCSRIDLLSSLIYVSQNSpecifications
RIPK1 (Human) Recombinant Protein (Q01) | |
Yes | |
37.84kDa | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |