missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RIN3 Partial ORF (NP_079108, 328 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_079108 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79890 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035655
|
Abnova™
H00079890-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045655
|
Abnova™
H00079890-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (see RAB5A; MIM 179512) (Kajiho et al., 2003 [PubMed 12972505]).[supplied by OMIM]
Sequence: GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTPSpecifications
NP_079108 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762H1613/FLJ11700/FLJ22439 | |
RIN3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79890 | |
RIN3 (Human) Recombinant Protein (Q01) | |
GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTP | |
RUO | |
RIN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |