Learn More
Abnova™ Human RHPN2 Full-length ORF (NP_149094.2, 1 a.a. - 686 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00085415-P02.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton. [provided by RefSeq]
Sequence: MTDALLPAAPQPLEKKNDGYFRKGCNPLAQTGRSKLQNQRAALNQQILKAVRMRTGAENLLKVATNSKVREQVRLELSFVNSDLQMLKEELEGLNISVGVYQNTEEAFTIPLIPLGLKETKDVDFAVVLKDFILEHYSEDGYLYEDEIADLMDLRQACRTPSRDEAGVELLMTYFIQLGFVESRFFPPTRQMGLLFTWYDSLTGVPVSQQNLLLEKASVLFNTGALYTQIGTRCDRQTQAGLESAIDAFQRAAGVLNYLKDTFTHTPSYDMSPAMLSVLVKMMLAQAQESVFEKISLPGIRNEFFMLVKVAQEAAKVGEVYQQLHAAMSQAPVKENIPYSWASLACVKAHHYAALAHYFTAILLIDHQVKPGTDLDHQEKCLSQLYDHMPEGLTPLATLKNDQQRRQLGKSHLRRAMAHHEESVREASLCKKLRSIEVLQKVLCAAQERSRLTYAQHQEEDDLLNLIDAPSVVAKTEQEVDIILPQFSKLTVTDFFQKLGPLSVFSANKRWTPPRSIRFTAEEGDLGFTLRGNAPVQVHFLDPYCSASVAGAREGDYIVSIQLVDCKWLTLSEVMKLLKSFGEDEIEMKVVSLLDSTSSMHNKSATYSVGMQKTYSMICLAIDDDDKTDKTKKISKKLSFLSWGTNKNRQKSASTLCLPSVGAARPQVKKKLPSPFSLLNSDSSWYSpecifications
NP_149094.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
103.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RhoBP/p76RBE | |
RHPN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
85415 | |
RHPN2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTDALLPAAPQPLEKKNDGYFRKGCNPLAQTGRSKLQNQRAALNQQILKAVRMRTGAENLLKVATNSKVREQVRLELSFVNSDLQMLKEELEGLNISVGVYQNTEEAFTIPLIPLGLKETKDVDFAVVLKDFILEHYSEDGYLYEDEIADLMDLRQACRTPSRDEAGVELLMTYFIQLGFVESRFFPPTRQMGLLFTWYDSLTGVPVSQQNLLLEKASVLFNTGALYTQIGTRCDRQTQAGLESAIDAFQRAAGVLNYLKDTFTHTPSYDMSPAMLSVLVKMMLAQAQESVFEKISLPGIRNEFFMLVKVAQEAAKVGEVYQQLHAAMSQAPVKENIPYSWASLACVKAHHYAALAHYFTAILLIDHQVKPGTDLDHQEKCLSQLYDHMPEGLTPLATLKNDQQRRQLGKSHLRRAMAHHEESVREASLCKKLRSIEVLQKVLCAAQERSRLTYAQHQEEDDLLNLIDAPSVVAKTEQEVDIILPQFSKLTVTDFFQKLGPLSVFSANKRWTPPRSIRFTAEEGDLGFTLRGNAPVQVHFLDPYCSASVAGAREGDYIVSIQLVDCKWLTLSEVMKLLKSFGEDEIEMKVVSLLDSTSSMHNKSATYSVGMQKTYSMICLAIDDDDKTDKTKKISKKLSFLSWGTNKNRQKSASTLCLPSVGAARPQVKKKLPSPFSLLNSDSSWY | |
RUO | |
RHPN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |