missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human REC8 Full-length ORF (AAH04159.1, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number AAH04159.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9985
Molecular Weight (g/mol) 89.1kDa
View More Specs

Prodotti 2
Codice del prodotto Marca Quantity Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantity Prezzo Quantità e disponibilità  
16184322
missing translation for 'viewDocuments' Dettagli della promozione
Abnova™
H00009985-P01.25UG
25 ug
508.00 €
25µg
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16174322
missing translation for 'viewDocuments' Dettagli della promozione
Abnova™
H00009985-P01.10UG
10 ug
335.00 €
10µg
Visualizzare prodotti alternativi
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

This gene encodes a member of the kleisin family of SMC (structural maintenance of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene. [provided by RefSeq]

Sequence: MFYYPNVLQRHTGCFATIWLAATRGSRLVKREYLRVNVVKTCEEILNYVLVRVQPPQPGLPRPRFSLYLSAQLQIGVIRVYSQQCQYLVEDIQHILERLHRAQLQIRIDMETELPSLLLPNHLAMMETLEDAPDPFFGMMSVDPRLPSPFDIPQIRHLLEAAIPERVEEIPPEVPTEPREPERIPVTVLPPEAITILEAEPIRMLEIEGERELPEVSRRELDLLIAEEEEAILLEIPRLPPPAPAEVEGIGEALGPEELRLTGWEPGALLMEVTPPEELRLPAPPSPERRPPVPPPPRRRRRRRLLFWDKETQISPEKFQEQLQTRAHCWECPMVQPPERTIRGPAELFRTPTLSGWLPPELLGLWTHCAQPPPKALRRELPEEAAAEEERRKIEVPSEIEVPREALEPSFPLMVSLEISLEAAEEEKSRISLIPPEERWAWPEVEAPEAPALPVVPELPEVPMEMPLVLPPELELLSLEAVHRAVALELQANREPDFSSLVSPLSPRRMAARVFYLLLVLSAQQILHVKQEKPYGRLLIQPGPRFH
Specifica

Specifica

AAH04159.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.1kDa
Glutathione Sepharose 4 Fast Flow
MFYYPNVLQRHTGCFATIWLAATRGSRLVKREYLRVNVVKTCEEILNYVLVRVQPPQPGLPRPRFSLYLSAQLQIGVIRVYSQQCQYLVEDIQHILERLHRAQLQIRIDMETELPSLLLPNHLAMMETLEDAPDPFFGMMSVDPRLPSPFDIPQIRHLLEAAIPERVEEIPPEVPTEPREPERIPVTVLPPEAITILEAEPIRMLEIEGERELPEVSRRELDLLIAEEEEAILLEIPRLPPPAPAEVEGIGEALGPEELRLTGWEPGALLMEVTPPEELRLPAPPSPERRPPVPPPPRRRRRRRLLFWDKETQISPEKFQEQLQTRAHCWECPMVQPPERTIRGPAELFRTPTLSGWLPPELLGLWTHCAQPPPKALRRELPEEAAAEEERRKIEVPSEIEVPREALEPSFPLMVSLEISLEAAEEEKSRISLIPPEERWAWPEVEAPEAPALPVVPELPEVPMEMPLVLPPELELLSLEAVHRAVALELQANREPDFSSLVSPLSPRRMAARVFYLLLVLSAQQILHVKQEKPYGRLLIQPGPRFH
RUO
REC8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
9985
REC8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HR21spB/MGC950/REC8L1/Rec8p
REC8
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati