missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBP7 Partial ORF (NP_443192.1, 35 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00116362-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101527

  • 341.70 € / 10µg

Estimated Shipment: 21-01-2022
Add to basket

Description

Description

Due to its chemical instability and low solubility in aqueous solution, vitamin A requires cellular retinol-binding proteins (CRBPs), such as RBP7, for stability, internalization, intercellular transfer, homeostasis, and metabolism.[supplied by OMIM]

Sequence: KLLKPQKVIEQNGDSFTIHTNSSLRNYFVKFKVGEEFDEDNRGLDNRKCKSLVIWDNDRLTCIQKGEKKNRGWTHWIEGDKLHLEMFCEGQVCKQTFQRA
Specifications

Specifications

NP_443192.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLLKPQKVIEQNGDSFTIHTNSSLRNYFVKFKVGEEFDEDNRGLDNRKCKSLVIWDNDRLTCIQKGEKKNRGWTHWIEGDKLHLEMFCEGQVCKQTFQRA
RUO
RBP7
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
116362
RBP7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRBP4/CRBPIV/MGC70641
RBP7
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions