missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBBP5 Full-length ORF (NP_005048.2, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005929-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100112

  • 341.70 € / 10µg

Estimated Shipment: 14-01-2022
Add to basket

Description

Description

The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. [provided by RefSeq]

Sequence: MNLELLESFGQNYPEEADGTLDCISMALTCTFNRWGTLLAVGCNDGRIVIWDFLTRGIAKIISAHIHPVCSLCWSRDGHKLVSASTDNIVSQWDVLSGDCDQRFRFPSPILKVQYHPRDQNKVLVCPMKSAPVMLTLSDSKHVVLPVDDDSDLNVVASFDRRGEYIYTGNAKGKILVLKTDSQDLVASFRVTTGTSNTTAIKSIEFARKGSCFLINTADRIIRVYDGREILTCGRDGEPEPMQKLQDLVNRTPWKKCCFSGDGEYIVAGSARQHALYIWEKSIGNLVKILHGTRGELLLDVAWHPVRPIIASISSGVVSIWAQNQVENWSAFAPDFKELDENVEYEERESEFDIEDEDKSEPEQTGADAAEDEEVDVTSVDPIAAFCSSDEELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKGKEKDSPFKPKLYKGDRGLPLEGSAKGKVQAELSQPLTAGGAISELL
Specifications

Specifications

NP_005048.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RBQ3/SWD1
RBBP5
Yes
wheat germ expression system
Liquid
5929
RBBP5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNLELLESFGQNYPEEADGTLDCISMALTCTFNRWGTLLAVGCNDGRIVIWDFLTRGIAKIISAHIHPVCSLCWSRDGHKLVSASTDNIVSQWDVLSGDCDQRFRFPSPILKVQYHPRDQNKVLVCPMKSAPVMLTLSDSKHVVLPVDDDSDLNVVASFDRRGEYIYTGNAKGKILVLKTDSQDLVASFRVTTGTSNTTAIKSIEFARKGSCFLINTADRIIRVYDGREILTCGRDGEPEPMQKLQDLVNRTPWKKCCFSGDGEYIVAGSARQHALYIWEKSIGNLVKILHGTRGELLLDVAWHPVRPIIASISSGVVSIWAQNQVENWSAFAPDFKELDENVEYEERESEFDIEDEDKSEPEQTGADAAEDEEVDVTSVDPIAAFCSSDEELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKGKEKDSPFKPKLYKGDRGLPLEGSAKGKVQAELSQPLTAGGAISELL
RUO
RBBP5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Promotions

Promotions