Learn More
Abnova™ Human RASGRF1 Partial ORF (NP_002882, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_002882 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5923 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184145
|
Abnova™
H00005923-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174145
|
Abnova™
H00005923-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. The studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]
Sequence: MQKGIRLNDGHVASLGLLARKDGTRKGYLSKRSSDNTKWQTKWFALLQNLLFYFESDSSSRPSGLYLLEGCVCDRAPSPKPALSAKEPLEKQHYFTVNFSHENQKALELRSpecifications
NP_002882 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC25/CDC25L/GNRP/GRF1/GRF55/H-GRF55/PP13187 | |
RASGRF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5923 | |
RASGRF1 (Human) Recombinant Protein (Q01) | |
MQKGIRLNDGHVASLGLLARKDGTRKGYLSKRSSDNTKWQTKWFALLQNLLFYFESDSSSRPSGLYLLEGCVCDRAPSPKPALSAKEPLEKQHYFTVNFSHENQKALELR | |
RUO | |
RASGRF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |