missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAB2B Partial ORF (NP_116235, 127 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084932-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100957

  • 508.00 € / 25µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking; see MIM 179508.[supplied by OMIM]

Sequence: RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCC
Specifications

Specifications

NP_116235
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCC
RUO
RAB2B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84932
RAB2B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14824
RAB2B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions