missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAB2B Partial ORF (NP_116235, 127 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_116235 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84932 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100957
|
Abnova™
H00084932-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190947
|
Abnova™
H00084932-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rab proteins are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking; see MIM 179508.[supplied by OMIM]
Sequence: RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCCSpecifications
NP_116235 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14824 | |
RAB2B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84932 | |
RAB2B (Human) Recombinant Protein (Q01) | |
RDVKREEGEAFAREHGLIFMETSAKTACNVEEAFINTAKEIYRKIQQGLFDVHNEANGIKIGPQQSISTSVGPSASQRNSRDIGSNSGCC | |
RUO | |
RAB2B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |