missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PVRL1 Partial ORF (NP_002846, 33 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00005818-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: VQVNDSMYGFIGTDVVLHCSFANPLPSVKITQVTWQKSTNGSKQNVAIYNPSMGVSVLAPYRERVEFLRPSFTDGTIRLSRLELEDEGVYICEFATFPTGNRESQLNLTVSpezifikation
NP_002846 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VQVNDSMYGFIGTDVVLHCSFANPLPSVKITQVTWQKSTNGSKQNVAIYNPSMGVSVLAPYRERVEFLRPSFTDGTIRLSRLELEDEGVYICEFATFPTGNRESQLNLTV | |
RUO | |
PVRL1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5818 | |
PVRL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD111|CLPED1|ED4|HIgR|HVEC|MGC142031|MGC16207|OFC7|PRR|PRR1|PVRR|PVRR1|SK-12|nectin-1 | |
PVRL1 | |
Yes |