missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTPRN2 Partial ORF (NP_002838, 108 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number NP_002838
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5799
Molecular Weight (g/mol) 37.29kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16183895
View Documents Promotion Details
Abnova™
H00005799-Q01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16173895
View Documents Promotion Details
Abnova™
H00005799-Q01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domain, and thus represents a receptor-type PTP. The catalytic domain of this PTP is most closely related to PTPRN/IA-2beta. This PTP and PTPRN are both found to be major autoantigens associated with insulin-dependent diabetes mellitus. Three alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported. [provided by RefSeq]

Sequence: ELADLPKTYLRRPEASSPARPSKHSVGSERRYSREGGAALANALRRHLPFLEALSQAPASDVLARTHTAQDRPPAEGDDRFSESILTYVAHTSALTYPPGPRTQL
Specifications

Specifications

NP_002838
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IA-2beta/IAR/ICAAR/PTPRP
PTPRN2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
5799
PTPRN2 (Human) Recombinant Protein (Q01)
ELADLPKTYLRRPEASSPARPSKHSVGSERRYSREGGAALANALRRHLPFLEALSQAPASDVLARTHTAQDRPPAEGDDRFSESILTYVAHTSALTYPPGPRTQL
RUO
PTPRN2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications