missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PSTPIP1 Partial ORF (NP_003969, 148 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009051-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139815

  • 508.00 € / 25µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene binds to the cytoplasmic tail of CD2, an effector of T cell activation and adhesion, negatively affecting CD2-triggered T cell activation. The encoded protein appears to be a scaffold protein and a regulator of the actin cytoskeleton. It has also been shown to bind ABL1, PTPN18, WAS, CD2AP, and PTPN12. Mutations in this gene are a cause of PAPA syndrome. [provided by RefSeq]

Sequence: CRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDE
Specifications

Specifications

NP_003969
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDE
RUO
PSTPIP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9051
PSTPIP1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD2BP1/CD2BP1L/CD2BP1S/H-PIP/PAPAS/PSTPIP
PSTPIP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions