missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPM1L Full-length ORF (NP_640338.2, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00151742-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138693

  • 508.00 € / 25µg
Estimated Shipment: 28-08-2024
to see stock.

Description

Description

PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK; see MIM 601158) signaling cascades that respond to extracellular stimuli (Jin et al., 2004 [PubMed 15560375]).[supplied by OMIM]

Sequence: MIEDTMTLLSLLGRIMRYFLLRPETLFLLCISLALWSYFFHTDEVKTIVKSSRDAVKMVKGKVAEIMQNDRLGGLDVLEAEFSKTWEFKNHNVAVYSIQGRRDHMEDRFEVLTDLANKTHPSIFGIFDGHGGETAAEYVKSRLPEALKQHLQDYEKDKENSVLSYQTILEQQILSIDREMLEKLTVSYDEAGTTCLIALLSDKDLTVANVGDSRGVLCDKDGNAIPLSHDHKPYQLKERKRIKRAGGFISFNGSWRVQGILAMSRSLGDYPLKNLNVVIPDPDILTFDLDKLQPEFMILASDGLWDAFSNEEAVRFIKERLDEPHFGAKSIVLQSFYRGCPDNITVMVVKFRNSSKTEEQ
Specifications

Specifications

NP_640338.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC132545/MGC132547/PP2C-epsilon/PP2CE/PPM1-LIKE
PPM1L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
151742
PPM1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIEDTMTLLSLLGRIMRYFLLRPETLFLLCISLALWSYFFHTDEVKTIVKSSRDAVKMVKGKVAEIMQNDRLGGLDVLEAEFSKTWEFKNHNVAVYSIQGRRDHMEDRFEVLTDLANKTHPSIFGIFDGHGGETAAEYVKSRLPEALKQHLQDYEKDKENSVLSYQTILEQQILSIDREMLEKLTVSYDEAGTTCLIALLSDKDLTVANVGDSRGVLCDKDGNAIPLSHDHKPYQLKERKRIKRAGGFISFNGSWRVQGILAMSRSLGDYPLKNLNVVIPDPDILTFDLDKLQPEFMILASDGLWDAFSNEEAVRFIKERLDEPHFGAKSIVLQSFYRGCPDNITVMVVKFRNSSKTEEQ
RUO
PPM1L
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions