missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PCSK7 Full-length ORF (AAH10696.1, 1 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009159-P02.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MPKGRQKVPHLDAPLGLPTCLWLELAGLFLLVPWVMGLAGTGGPDGQGTGGPSWAVHLESLEGDGEEETLEQQADALAQAAGLVNAGRIGELQGHYLFVQPAGHRPALEVEAIRQQVEAVLAGHEAVRWHSEQRLLRRAKRSVHFNDPKYPQQWHLNNRRSPGRDINVTGVWERNVTGRGVTVVVVDDGVEHTIQDIAPNYSPEGSYDLNSNDPDPMPHPDVENGNHHGTRCAGEIAAVPNNSFCAVGVAYGSRIAGIRVLDGPLTDSMEAVAFNKHYQINDIYSCSWGPDDDGKTVDGPHQLGKAALQHGVIAGRQGFGSIFVVASGNGGQHNDNCNYDGYANSIYTVTIGAVDEEGRMPFYAEECASMLAVTFSGGDKMLRSIVTTDWDLQKGTGCTEGHTGTSAAAPLAAGMIALMLQVRPCLTWRDVQHIIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLELKLFCPSGMMSLIGAPRSMDSDPNGFNDWTFSTVRCWGERARGTYRLVIRDVGDESFQVGILRQWQLTLYGSVWSAVDIRDRQRLLESAMSGKYLHDDFALPCPPGLKIPEEDGYTITPNTLKTLVLVGCFTVFWTVYYMLEVYLSQRNVASNQVCRSGPCHWPHRSRKAKEEGTELESVPLCSSKDPDEVETESRGPPTTSDLLAPDLLEQGDWSLSQNKSALDCPHQHLDVPHGKEEQICSpezifikation
AAH10696.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
111.87kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LPC|PC7|PC8|SPC7 | |
PCSK7 | |
Yes |
wheat germ expression system | |
Liquid | |
9159 | |
PCSK7 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPKGRQKVPHLDAPLGLPTCLWLELAGLFLLVPWVMGLAGTGGPDGQGTGGPSWAVHLESLEGDGEEETLEQQADALAQAAGLVNAGRIGELQGHYLFVQPAGHRPALEVEAIRQQVEAVLAGHEAVRWHSEQRLLRRAKRSVHFNDPKYPQQWHLNNRRSPGRDINVTGVWERNVTGRGVTVVVVDDGVEHTIQDIAPNYSPEGSYDLNSNDPDPMPHPDVENGNHHGTRCAGEIAAVPNNSFCAVGVAYGSRIAGIRVLDGPLTDSMEAVAFNKHYQINDIYSCSWGPDDDGKTVDGPHQLGKAALQHGVIAGRQGFGSIFVVASGNGGQHNDNCNYDGYANSIYTVTIGAVDEEGRMPFYAEECASMLAVTFSGGDKMLRSIVTTDWDLQKGTGCTEGHTGTSAAAPLAAGMIALMLQVRPCLTWRDVQHIIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLELKLFCPSGMMSLIGAPRSMDSDPNGFNDWTFSTVRCWGERARGTYRLVIRDVGDESFQVGILRQWQLTLYGSVWSAVDIRDRQRLLESAMSGKYLHDDFALPCPPGLKIPEEDGYTITPNTLKTLVLVGCFTVFWTVYYMLEVYLSQRNVASNQVCRSGPCHWPHRSRKAKEEGTELESVPLCSSKDPDEVETESRGPPTTSDLLAPDLLEQGDWSLSQNKSALDCPHQHLDVPHGKEEQIC | |
RUO | |
PCSK7 | |
Wheat Germ (in vitro) | |
GST |