missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHB11 Partial ORF (NP_061754, 27 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number NP_061754
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 56125
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16172083
View Documents Promotion Details
Abnova™
H00056125-Q01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16182083
View Documents Promotion Details
Abnova™
H00056125-Q01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: GSETWSFSVAEEMQSGSFVGNLAKDLGLKVRELSSRGARVVSNDKKQRLQLDINTGDLLLSETLDREELCGSIEPCVLHLQVLMQNPTQFLQIELQVRDI
Specifications

Specifications

NP_061754
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ME2/MGC138337/MGC142171/PCDH-BETA11
PCDHB11
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
56125
PCDHB11 (Human) Recombinant Protein (Q01)
GSETWSFSVAEEMQSGSFVGNLAKDLGLKVRELSSRGARVVSNDKKQRLQLDINTGDLLLSETLDREELCGSIEPCVLHLQVLMQNPTQFLQIELQVRDI
RUO
PCDHB11
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications