missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PAPSS1 Partial ORF (NP_005434, 144 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_005434 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9061 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16129845
|
Abnova™
H00009061-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139845
|
Abnova™
H00009061-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 [PubMed 10679223]). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2 (MIM 603005).[supplied by OMIM]
Sequence: QIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAESpecifications
NP_005434 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATPSK1/PAPSS/SK1 | |
PAPSS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9061 | |
PAPSS1 (Human) Recombinant Protein (Q01) | |
QIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAE | |
RUO | |
PAPSS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |