Learn More
Abnova™ Human PANK1 Full-length ORF (NP_683879.1, 1 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00053354-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. Alternative splicing has been observed at this locus and three variants, each encoding a distinct isoform, have been identified. [provided by RefSeq]
Sequence: MKLINGKKQTFPWFGMDIGGTLVKLVYFEPKDITAEEEQEEVENLKSIRKYLTSNTAYGKTGIRDVHLELKNLTMCGRKGNLHFIRFPSCAMHRFIQMGSEKNFSSLHTTLCATGGGAFKFEEDFRMIADLQLHKLDELDCLIQGLLYVDSVGFNGKPECYYFENPTNPELCQKKPYCLDNPYPMLLVNMGSGVSILAVYSKDNYKRVTGTSLGGGTFLGLCCLLTGCETFEEALEMAAKGDSTNVDKLVKDIYGGDYERFGLQGSAVASSFGNMMSKEKRDSISKEDLARATLVTITNNIGSIARMCALNENIDRVVFVGNFLRINMVSMKLLAYAMDFWSKGQLKALFLEHEGYFGAVGALLELFKMTDDKSpecifications
NP_683879.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC24596/PANK/PANK1a/PANK1b | |
PANK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53354 | |
PANK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLINGKKQTFPWFGMDIGGTLVKLVYFEPKDITAEEEQEEVENLKSIRKYLTSNTAYGKTGIRDVHLELKNLTMCGRKGNLHFIRFPSCAMHRFIQMGSEKNFSSLHTTLCATGGGAFKFEEDFRMIADLQLHKLDELDCLIQGLLYVDSVGFNGKPECYYFENPTNPELCQKKPYCLDNPYPMLLVNMGSGVSILAVYSKDNYKRVTGTSLGGGTFLGLCCLLTGCETFEEALEMAAKGDSTNVDKLVKDIYGGDYERFGLQGSAVASSFGNMMSKEKRDSISKEDLARATLVTITNNIGSIARMCALNENIDRVVFVGNFLRINMVSMKLLAYAMDFWSKGQLKALFLEHEGYFGAVGALLELFKMTDDK | |
RUO | |
PANK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |