missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OTOP1 Partial ORF (NP_819056.1, 415 a.a. - 504 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00133060-Q01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16181827

  • 310.00 € / 10 Mikrogramm


Verfügbar ab
15-02-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: AEGHPRYTWYNLPYSILAIVEKYIQNLFIFESIHREPEKLSEDIQTLRVVTVCNGNTMPLASSCPKSGGVARDVAPQGKDMPPAANGNVC
Spezifikation

Spezifikation

NP_819056.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AEGHPRYTWYNLPYSILAIVEKYIQNLFIFESIHREPEKLSEDIQTLRVVTVCNGNTMPLASSCPKSGGVARDVAPQGKDMPPAANGNVC
RUO
OTOP1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
133060
OTOP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC163302|MGC163304
OTOP1
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation