missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL7 Partial ORF (NP_665741.1, 742 a.a. - 842 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_665741.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 114881 |
Molecular Weight (g/mol) | 36.85kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181437
|
Abnova™
H00114881-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171437
|
Abnova™
H00114881-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified. [provided by RefSeq]
Sequence: ELNELTAELKRSLPSTDTRLRPDQRYLEEGNIQAAEAQKRRIEQLQRDRRKVMEENNIVHQARFFRRQTDSSGKEWWVTNNTYWRLRAEPGYGNMDGAVLWSpecifications
NP_665741.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC71150/ORP7 | |
OSBPL7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
114881 | |
OSBPL7 (Human) Recombinant Protein (Q01) | |
ELNELTAELKRSLPSTDTRLRPDQRYLEEGNIQAAEAQKRRIEQLQRDRRKVMEENNIVHQARFFRRQTDSSGKEWWVTNNTYWRLRAEPGYGNMDGAVLW | |
RUO | |
OSBPL7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |