missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL3 Partial ORF (NP_056365.1, 788 a.a. - 887 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_056365.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26031 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185266
|
Abnova™
H00026031-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175266
|
Abnova™
H00026031-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Several transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: FALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLWSpecifications
NP_056365.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667P1518/KIAA0704/MGC21526/ORP-3/ORP3/OSBP3 | |
OSBPL3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26031 | |
OSBPL3 (Human) Recombinant Protein (Q01) | |
FALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW | |
RUO | |
OSBPL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |