missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL3 Full-length ORF (NP_663162.1, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026031-P01.25ug

 View more versions of this product

Product Code. 16138022

  • 521.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Several transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW
Specifications

Specifications

NP_663162.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
120.2kDa
Glutathione Sepharose 4 Fast Flow
25 μg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp667P1518/KIAA0704/MGC21526/ORP-3/ORP3/OSBP3
OSBPL3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26031
OSBPL3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW
RUO
OSBPL3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.