missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NOS1AP Partial ORF (NP_055512.1, 99 a.a. - 202 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009722-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101216

  • 335.00 € / 10µg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: WTWDESKMLVMQDPIYRIFYVSHDSQDLKIFSYIARDGASNIFRCNVFKSKKKSQAMRIVRTVGQAFEVCHKLSLQHTQQNADGQEDGESERNSNSSGDPGRQL
Specifications

Specifications

NP_055512.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WTWDESKMLVMQDPIYRIFYVSHDSQDLKIFSYIARDGASNIFRCNVFKSKKKSQAMRIVRTVGQAFEVCHKLSLQHTQQNADGQEDGESERNSNSSGDPGRQL
RUO
NOS1AP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9722
NOS1AP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
6330408P19Rik/CAPON/MGC138500
NOS1AP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions